Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Platinum on alumina
Synonym(s):
Pt/Al
2
O
3
Products
Product Category
catalysts (3)
Brand
Sigma-Aldrich (5)
Sustainability
Feature
Physical Form
Quality Segment
Reagent Type
Core
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
205974
Platinum on alumina,
extent of labeling: 5 wt. % loading, powder
Expand
View Pricing
232114
Platinum on alumina,
extent of labeling: 1 wt. % loading, 3.2 mm, pellets
Expand
View Pricing
206016
Platinum on alumina,
extent of labeling: 0.5 wt. % loading, 3.2 mm, pellets
Expand
View Pricing
311324
Platinum on alumina,
extent of labeling: 5 wt. % loading, -325 mesh, powder
Expand
View Pricing
520705
Platinum on alumina,
extent of labeling: 0.5 wt. % loading, spheres
Expand
View Pricing
205966
Platinum on alumina,
extent of labeling: 1 wt. % loading, powder
Expand
View Pricing