Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Pipette tips for Velocity, robotic
Synonym(s):
Pipette tips for Velocity, robotic
Products
Product Category
pipette tips (5)
Manufacturer
Sterility
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
AXYVT250RLAB
Pipette tips for Velocity, robotic,
size 250 μL, pkg of 5x960tips/cs, racked in bottom sealed rack
Expand
View Pricing
AXYVT250RS
Pipette tips for Velocity, robotic,
size 250 μL, pkg of 5x960tips/cs, racked, clear, sterile
Expand
View Pricing
AXYVT250R
Pipette tips for Velocity, robotic,
size 250 μL, pkg of 5x960tips/cs, racked
Expand
View Pricing
AXYVT250RLABS
Pipette tips for Velocity, robotic,
size 250 μL, pkg of 5x960tips/cs, racked in bottom sealed rack, sterile
Expand
View Pricing
AXYVT20R
Pipette tips for Velocity, robotic,
size 20 μL, pkg of 5x960tips/cs, racked
Expand
View Pricing