Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Phosphodiesterase I from
Crotalus adamanteus
venom
Synonym(s):
5′-Exonuclease, Oligonucleate 5′-nucleotidohydrolase
CAS Number:
9025-82-5
EC Number:
232-806-5
Products
Brand
Sigma-Aldrich (2)
Biological Source
Physical Form
Tissue
Quality Segment
Package Size
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
P3243
Phosphodiesterase I from
Crotalus adamanteus
venom,
vial of ≥100 units, Purified
Expand
View Pricing
P3134
Phosphodiesterase I from
Crotalus adamanteus
venom,
Type VI, crude dried venom
Expand
View Pricing