Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Phloridzin dihydrate
Synonym(s):
1-[2-(β-D-Glucopyranosyloxy)-4,6-dihydroxyphenyl]-3-(4-hydroxyphenyl)-1-propanone, Phloretin 2′-β-D-glucopyranoside, Phloretin 2′-β-D-glucoside, Phlorizin dihydrate
Empirical Formula (Hill Notation):
C
21
H
24
O
10
· 2H
2
O
CAS Number:
7061-54-3
Molecular Weight:
472.44
EC Number:
200-487-1
Products
Product Category
bioactive small molecules (1)
building blocks (1)
carbohydrates (1)
Brand
Sigma-Aldrich (2)
Biological Source
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Optical Isomer
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
274313
Phloridzin dihydrate,
99%
Expand
View Pricing
P3449
Phloridzin dihydrate,
from apple wood, ≥99% (HPLC)
Expand
View Pricing