Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Phenylmagnesium bromide solution
Linear Formula:
C
6
H
5
MgBr
CAS Number:
100-58-3
Molecular Weight:
181.31
Products
Brand
Sigma-Aldrich (1)
Sustainability
Feature
Formula Weight
Physical Form
Quality Segment
Markush Class
Markush Group
Reaction Type
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
331376
Phenylmagnesium bromide solution,
1.0 M in THF
Expand
View Pricing
171565
Phenylmagnesium bromide solution,
3.0 M in diethyl ether
Expand
View Pricing
703575
Phenylmagnesium bromide solution,
2.9 M in 2-methyltetrahydrofuran
Expand
View Pricing