Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
PEEK Tubing
Products
Product Category
tubing (7)
Color
Special Grade
Natural (2)
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
Z226661
PEEK Tubing,
L × O.D. × I.D. 10 ft × 1/16 in. × 0.010 in., blue, max. pressure 6000 psi
Expand
View Pricing
Z227307
PEEK Tubing,
L × O.D. × I.D. 10 ft × 1/16 in. × 0.005 in., red, max. pressure 8000
Expand
View Pricing
54995
PEEK Tubing,
L × O.D. × I.D. 10 ft × 1/8 in. × 0.062 in., natural
Expand
View Pricing
Z226955
PEEK Tubing,
L × O.D. × I.D. 10 ft × 1/16 in. × 0.030 in., green, max. pressure 5000 psi
Expand
View Pricing
Z226688
PEEK Tubing,
L × O.D. × I.D. 10 ft × 1/16 in. × 0.007 in., yellow, max. pressure 8000
Expand
View Pricing
Z227293
PEEK Tubing,
L × O.D. × I.D. 10 ft × 1/16 in. × 0.020 in., orange, max. pressure 6000 psi
Expand
View Pricing
54994
PEEK Tubing,
L × O.D. × I.D. 10 ft × 1/16 in. × 0.055 in., natural, max. pressure 1000 psi
Expand
View Pricing