Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
PCR microplate for Roche
®
480 LightCycler
®
Products
Product Category
plates (4)
Color
Manufacturer
Binding Type
Volume (µL)
Wells
Working Volume (ml)
Color
Package Size
Sterility
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
AXYPCR96LC480W
PCR microplate for Roche
®
480 LightCycler
®
,
with UC-500 film, size 96 wells, pkg of 5x10plates/cs, white
Expand
View Pricing
AXYPCR96LC480WNF
PCR microplate for Roche
®
480 LightCycler
®
,
size 96 wells, white, no sealing film
Expand
View Pricing
AXYPCR384LC480W
PCR microplate for Roche
®
480 LightCycler
®
,
with film, white, size 384 wells, pkg of 5x10plates/cs, conical bottom white polypropylene wells
Expand
View Pricing
AXYPCR384LC480WNF
PCR microplate for Roche
®
480 LightCycler
®
,
no film, size 384 wells, white, pkg of 5x10plates/cs
Expand
View Pricing