Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Palladium on calcium carbonate
Linear Formula:
Pd
Products
Product Category
catalysts (1)
Brand
Sigma-Aldrich (3)
Sustainability
Feature
Physical Form
Reaction Type
Reagent Type
Core
Special Grade
Puriss (1)
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
205737
Palladium on calcium carbonate,
extent of labeling: 5 wt. % loading, poisoned with lead
Expand
View Pricing
76030
Palladium on calcium carbonate,
~10% Pd basis
Expand
View Pricing
214353
Palladium on calcium carbonate,
extent of labeling: 5 wt. % loading, unreduced
Expand
View Pricing
908002
Palladium on calcium carbonate,
Evonik Noblyst
®
P1152 5% Pd
Expand
View Pricing