Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Palladium(II) chloride
Synonym(s):
Palladium(II) chloride (59% Pd)
Empirical Formula (Hill Notation):
Cl
2
Pd
CAS Number:
7647-10-1
Molecular Weight:
177.33
EC Number:
231-596-2
Products
Brand
Sigma-Aldrich (7)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Chemical Composition
Quality Segment
Reaction Type
Reagent Type
Core
Special Grade
Anhydrous (2)
Purum (1)
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
520659
Palladium(II) chloride,
≥99.9%
Expand
View Pricing
205885
Palladium(II) chloride,
ReagentPlus
®
, 99%
Expand
View Pricing
76050
Palladium(II) chloride,
anhydrous, 60% Pd basis
Expand
View Pricing
8.07110
Palladium(II) chloride,
59% Pd, anhydrous, for synthesis
Expand
View Pricing
323373
Palladium(II) chloride,
99.995%
Expand
View Pricing
921033
Palladium(II) chloride,
≥99.99% trace metals basis
Expand
View Pricing
14814
Palladium(II) chloride,
anhydrous, 59-60% palladium (Pd) basis
Expand
View Pricing