Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Palladium hydroxide on carbon
Synonym(s):
Pearlman’s catalyst
Empirical Formula (Hill Notation):
H
2
O
2
Pd
CAS Number:
12135-22-7
Molecular Weight:
140.43
EC Number:
235-219-2
Products
Product Category
catalysts (2)
Formula Weight
Physical Form
Quality Segment
Reaction Type
Reagent Type
Core
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
212911
Palladium hydroxide on carbon,
extent of labeling: 20 wt. % loading (dry basis), matrix carbon, wet support
Expand
View Pricing
330094
Palladium hydroxide on carbon,
extent of labeling: 20 wt. % loading (dry basis), matrix carbon, wet support
Expand
View Pricing