Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Orange G
Synonym(s):
1-Phenylazo-2-naphthol-6,8-disulfonic acid disodium salt, 7-Hydroxy-8-phenylazo-1,3-naphthalenedisulfonic acid disodium salt, Acid Orange 10, Wool Orange 2G
Empirical Formula (Hill Notation):
C
16
H
10
N
2
Na
2
O
7
S
2
CAS Number:
1936-15-8
Molecular Weight:
452.37
EC Number:
217-705-6
Products
Product Category
stains (2)
Brand
Sigma-Aldrich (3)
Color
Formula Weight
Physical Form
Chemical Composition
Quality Segment
Application
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
O3756
Orange G,
for NA electrophoresis
Expand
View Pricing
O7252
Orange G,
Dye content, ≥80%, certified by the Biological Stain Commission, powder
Expand
View Pricing
861286
Orange G,
certified by the Biological Stain Commission, Dye content 80 %
Expand
View Pricing