Skip to Content
Merck
(−)-O,O′-Dibenzoyl-L-tartaric acid mono(dimethylamide) ≥99.0%

(−)-O,O′-Dibenzoyl-L-tartaric acid mono(dimethylamide)

(−)-O,O′-Dibenzoyl-L-tartaric acid mono(dimethylamide) ≥99.0%
Empirical Formula (Hill Notation):
C20H19NO7
CAS Number:
Molecular Weight:
385.37

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing