Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Ohaus
®
Navigator
®
Portable Balance
Products
Product Category
balances (18)
Manufacturer
Length (cm)
Weighing Capacity (g)
Resolution (g)
Package Size
Filter Products
Showing 1-18 of 18
Compare
Product Number
Description
Pricing
OH30456385
Ohaus
®
Navigator
®
Portable Balance,
Model NV221, weighing capacity 220 g, resolution: 0.1 g, AC/DC input 230 V, universal plug set
Expand
View Pricing
OH30456387
Ohaus
®
Navigator
®
Portable Balance,
Model NV1201, weighing capacity 1200 g, resolution: 0.1 g, AC/DC input 230 V, universal plug set
Expand
View Pricing
OH30456384
Ohaus
®
Navigator
®
Portable Balance,
Model NV622, weighing capacity 620 g, resolution: 0.01 g, AC/DC input 230 V, universal plug set
Expand
View Pricing
OH30456386
Ohaus
®
Navigator
®
Portable Balance,
Model NV621, weighing capacity 620 g, resolution: 0.1 g, AC/DC input 230 V, universal plug set
Expand
View Pricing
OH30456388
Ohaus
®
Navigator
®
Portable Balance,
Model NV2201, weighing capacity 2200 g, resolution: 0.1 g, AC/DC input 230 V, universal plug set
Expand
View Pricing
OH30456392
Ohaus
®
Navigator
®
Portable Balance,
Model NVT2200, weighing capacity 2200 g, resolution: 1 g, AC/DC input 230 V, universal plug set
Expand
View Pricing
OH30456394
Ohaus
®
Navigator
®
Portable Balance,
Model NVT12000, weighing capacity 12000 g, resolution: 1 g, AC/DC input 230 V, universal plug set
Expand
View Pricing
OH30456393
Ohaus
®
Navigator
®
Portable Balance,
Model NVT6200, weighing capacity 6200 g, resolution: 1 g, AC/DC input 230 V, universal plug set
Expand
View Pricing
OH30456382
Ohaus
®
Navigator
®
Portable Balance,
Model NV222, weighing capacity 220 g, resolution: 0.01 g, AC/DC input 230 V, universal plug set
Expand
View Pricing
OH30467838
Ohaus
®
Navigator
®
Portable Balance,
Model NVT16000M, weighing capacity 16000 g, resolution: 5 g, AC/DC input 230 V, universal plug set
Expand
View Pricing
OH30456391
Ohaus
®
Navigator
®
Portable Balance,
Model NVT6201, weighing capacity 6200 g, resolution: 0.1 g, AC/DC input 230 V, universal plug set
Expand
View Pricing
OH30467836
Ohaus
®
Navigator
®
Portable Balance,
Model NVT3200M, weighing capacity 3200 g, resolution: 1 g, AC/DC input 230 V, universal plug set
Expand
View Pricing
OH30456395
Ohaus
®
Navigator
®
Portable Balance,
Model NVT22000, weighing capacity 22000 g, resolution: 1 g, AC/DC input 230 V, universal plug set
Expand
View Pricing
OH30456383
Ohaus
®
Navigator
®
Portable Balance,
Model NV422, weighing capacity 420 g, resolution: 0.01 g, AC/DC input 230 V, universal plug set
Expand
View Pricing
OH30467835
Ohaus
®
Navigator
®
Portable Balance,
Model NVT1601M, weighing capacity 1600 g, resolution: 0.5 g, AC/DC input 230 V, universal plug set
Expand
View Pricing
OH30456390
Ohaus
®
Navigator
®
Portable Balance,
Model NVT4201, weighing capacity 4200 g, resolution: 0.1 g, AC/DC input 230 V, universal plug set
Expand
View Pricing
OH30456389
Ohaus
®
Navigator
®
Portable Balance,
Model NVT2201, weighing capacity 2200 g, resolution: 0.1 g, AC/DC input 230 V, universal plug set
Expand
View Pricing
OH30467837
Ohaus
®
Navigator
®
Portable Balance,
Model NVT6400M, weighing capacity 6400 g, resolution: 2 g, AC/DC input 230 V, universal plug set
Expand
View Pricing