Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Ohaus
®
Explorer
®
precision balance
Products
Product Category
balances (76)
Manufacturer
Length (cm)
Weighing Capacity (g)
Resolution (g)
Package Size
Filter Products
Showing 1-20 of 76
Page 1 of 4
Page 1 of 4
Compare
Product Number
Description
Pricing
Z760536
Ohaus
®
Explorer
®
precision balance,
Model EX423, weighing capacity 420 g, precision: 0.001 g, AC/DC input 230 V AC (universal plugs)
Expand
View Pricing
Z760501
Ohaus
®
Explorer
®
precision balance,
Model EX223, weighing capacity 220 g, precision: 0.001 g, AC/DC input 230 V AC (universal plugs)
Expand
View Pricing
Z743921
Ohaus
®
Explorer
®
precision balance,
Model EX223/E, weighing capacity 220 g, Precision 1 mg, AC/DC input 230 V (Euro Plug and UK Plug)
Expand
View Pricing
OH30061979GB
Ohaus
®
Explorer
®
precision balance,
Model EX223/E, weighing capacity 220 g, resolution: 0.001 g, AC/DC input 220/240 V, UK plug
Expand
View Pricing
Z760595
Ohaus
®
Explorer
®
precision balance,
Model EX2202, weighing capacity 2200 g, precision: 0.01 g, AC/DC input 230 V AC (universal plugs)
Expand
View Pricing
Z743923
Ohaus
®
Explorer
®
precision balance,
Model EX423/E, weighing capacity 420 g, Precision 1 mg, AC/DC input 230 V (Euro Plug and UK Plug)
Expand
View Pricing
OH83021365GB
Ohaus
®
Explorer
®
precision balance,
Model EX10201, weighing capacity 10200 g, resolution: 0.1 g, AC/DC input 220/240 V, UK plug
Expand
View Pricing
OH30061977GB
Ohaus
®
Explorer
®
precision balance,
Model EX224/AD, weighing capacity 220 g, resolution: 0.1 mg, AC/DC input 220/240 V, UK plug
Expand
View Pricing
Z760552
Ohaus
®
Explorer
®
precision balance,
Model EX623, weighing capacity 620 g, precision: 0.001 g, AC/DC input 230 V AC (universal plugs)
Expand
View Pricing
Z760617
Ohaus
®
Explorer
®
precision balance,
Model EX4202, weighing capacity 4200 g, precision: 0.01 g, AC/DC input 230 V AC (universal plugs)
Expand
View Pricing
OH30078077
Ohaus
®
Explorer
®
precision balance,
Model EX35001-GB, weighing capacity 35000 g, resolution: 0.1 g, AC/DC input 210 - 2310 V, UK plug
Expand
View Pricing
Z760579
Ohaus
®
Explorer
®
precision balance,
Model EX1103, weighing capacity 1100 g, precision: 0.001 g, AC/DC input 230 V AC (universal plugs)
Expand
View Pricing
Z760633
Ohaus
®
Explorer
®
precision balance,
Model EX6202, weighing capacity 6200 g, precision: 0.01 g, AC/DC input 230 V AC (universal plugs)
Expand
View Pricing
Z760706
Ohaus
®
Explorer
®
precision balance,
Model EX10201, weighing capacity 10200 g, precision: 0.1 g, AC/DC input 230 V AC (universal plugs)
Expand
View Pricing
OH83021356GB
Ohaus
®
Explorer
®
precision balance,
Model EX2202, weighing capacity 2200 g, resolution: 0.01 g, AC/DC input 220/240 V, UK plug
Expand
View Pricing
OH30061984GB
Ohaus
®
Explorer
®
precision balance,
Model EX6201/E, weighing capacity 6200 g, resolution: 0.1 g, AC/DC input 220/240 V, UK plug
Expand
View Pricing
Z760668
Ohaus
®
Explorer
®
precision balance,
Model EX10202, weighing capacity 10200 g, precision: 0.01 g, AC/DC input 230 V AC (universal plugs)
Expand
View Pricing
OH83021362GB
Ohaus
®
Explorer
®
precision balance,
Model EX10202, weighing capacity 10200 g, resolution: 0.01 g, AC/DC input 220/240 V, UK plug
Expand
View Pricing
OH30078073
Ohaus
®
Explorer
®
precision balance,
Model EX24001-EU, weighing capacity 24000 g, resolution: 0.1 g, AC/DC input 210 - 2310 V, Schuko plug
Expand
View Pricing
OH83021343EU
Ohaus
®
Explorer
®
precision balance,
Model EX423M, weighing capacity 420 g, resolution: 0.001 g, AC/DC input 230 V, Schuko plug
Expand
View Pricing
Page 1 of 4
Page 1 of 4