Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Ohaus
®
Explorer
®
analytical balance
Products
Product Category
balances (15)
Manufacturer
Diameter (mm)
Weighing Capacity (g)
Package Size
Filter Products
Showing 1-15 of 15
Compare
Product Number
Description
Pricing
Z760455
Ohaus
®
Explorer
®
analytical balance,
Model EX224, weighing capacity 220 g, precision: 0.1 mg, AC/DC input 230 V AC (universal plugs)
Expand
View Pricing
Z760439
Ohaus
®
Explorer
®
analytical balance,
Model EX124, weighing capacity 120 g, precision: 0.1 mg, AC/DC input 230 V AC (universal plugs)
Expand
View Pricing
Z760471
Ohaus
®
Explorer
®
analytical balance,
Model EX324, weighing capacity 320 g, precision: 0.1 mg, AC/DC input 230 V AC (universal plugs)
Expand
View Pricing
OH83021336GB
Ohaus
®
Explorer
®
analytical balance,
Model EX324M, weighing capacity 320 g, resolution: 0.1 mg, AC/DC input 220/240 V, UK plug
Expand
View Pricing
Z743918
Ohaus
®
Explorer
®
analytical balance,
Model EX224M, weighing capacity 220 g, Precision 0.1 mg, AC/DC input 230 V (Euro Plug and UK Plug)
Expand
View Pricing
OH83021332GB
Ohaus
®
Explorer
®
analytical balance,
Model EX224, weighing capacity 220 g, resolution: 0.1 mg, AC/DC input 220/240 V, UK plug
Expand
View Pricing
OH83021335GB
Ohaus
®
Explorer
®
analytical balance,
Model EX324, weighing capacity 320 g, resolution: 0.1 mg, AC/DC input 220/240 V, UK plug
Expand
View Pricing
OH83021331GB
Ohaus
®
Explorer
®
analytical balance,
Model EX124, weighing capacity 120 g, resolution: 0.1 mg, AC/DC input 220/240 V, UK plug
Expand
View Pricing
OH83021332EU
Ohaus
®
Explorer
®
analytical balance,
Model EX224, weighing capacity 220 g, resolution: 0.1 mg, AC/DC input 230 V, Schuko plug
Expand
View Pricing
Z743919
Ohaus
®
Explorer
®
analytical balance,
Model EX324M, weighing capacity 320 g, Precision 0.1 mg, AC/DC input 230 V (Euro Plug and UK Plug)
Expand
View Pricing
OH83021331EU
Ohaus
®
Explorer
®
analytical balance,
Model EX124, weighing capacity 120 g, resolution: 0.1 mg, AC/DC input 230 V, Schuko plug
Expand
View Pricing
OH83021335EU
Ohaus
®
Explorer
®
analytical balance,
Model EX324, weighing capacity 320 g, resolution: 0.1 mg, AC/DC input 230 V, Schuko plug
Expand
View Pricing
OH83021333GB
Ohaus
®
Explorer
®
analytical balance,
Model EX224M, weighing capacity 220 g, resolution: 0.1 mg, AC/DC input 220/240 V, UK plug
Expand
View Pricing
OH83021336EU
Ohaus
®
Explorer
®
analytical balance,
Model EX324M, weighing capacity 320 g, resolution: 0.1 mg, AC/DC input 230 V, Schuko plug
Expand
View Pricing
OH83021333EU
Ohaus
®
Explorer
®
analytical balance,
Model EX224M, weighing capacity 220 g, resolution: 0.1 mg, AC/DC input 230 V, Schuko plug
Expand
View Pricing