Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Ohaus
®
Adventurer
®
Precision Balance
Products
Product Category
balances (30)
Manufacturer
Length (cm)
Weighing Capacity (g)
Resolution (g)
Package Size
Filter Products
Showing 1-20 of 30
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
OH30127961
Ohaus
®
Adventurer
®
Precision Balance,
Model AX223/E, weighing capacity 220 g, resolution: 0.001 g, AC/DC input 210 - 2310 V, universal plug set (UK/EU Plugset)
Expand
View Pricing
OH30127963
Ohaus
®
Adventurer
®
Precision Balance,
Model AX423/E, weighing capacity 420 g, resolution: 0.001 g, AC/DC input 210 - 2310 V, universal plug set (UK/EU Plugset)
Expand
View Pricing
OH30122617
Ohaus
®
Adventurer
®
Precision Balance,
Model AX4202/E, weighing capacity 4200 g, resolution: 0.01 g, AC/DC input 210 - 2310 V, universal plug set (UK/EU Plugset)
Expand
View Pricing
OH30122620
Ohaus
®
Adventurer
®
Precision Balance,
Model AX8201/E, weighing capacity 8200 g, resolution: 0.1 g, AC/DC input 210 - 2310 V, universal plug set (UK/EU Plugset)
Expand
View Pricing
OH30127968
Ohaus
®
Adventurer
®
Precision Balance,
Model AX2202, weighing capacity 2200 g, resolution: 0.01 g, AC/DC input 230 V, universal plug set
Expand
View Pricing
OH30122615
Ohaus
®
Adventurer
®
Precision Balance,
Model AX523/E, weighing capacity 520 g, resolution: 0.001 g, AC/DC input 210 - 2310 V, universal plug set (UK/EU Plugset)
Expand
View Pricing
OH30122614
Ohaus
®
Adventurer
®
Precision Balance,
Model AX523, weighing capacity 520 g, resolution: 0.001 g, AC/DC input 210 - 2310 V, universal plug set (UK/EU Plugset)
Expand
View Pricing
OH30122618
Ohaus
®
Adventurer
®
Precision Balance,
Model AX5202, weighing capacity 5200 g, resolution: 0.01 g, AC/DC input 210 - 2310 V, universal plug set (UK/EU Plugset)
Expand
View Pricing
OH30127969
Ohaus
®
Adventurer
®
Precision Balance,
Model AX2202/E, weighing capacity 2200 g, resolution: 0.01 g, AC/DC input 230 V, universal plug set
Expand
View Pricing
OH30127960
Ohaus
®
Adventurer
®
Precision Balance,
Model AX223, weighing capacity 220 g, resolution: 0.001 g, AC/DC input 210 - 2310 V, universal plug set (UK/EU Plugset)
Expand
View Pricing
OH30122619
Ohaus
®
Adventurer
®
Precision Balance,
Model AX8201, weighing capacity 8200 g, resolution: 0.1 g, AC/DC input 210 - 2310 V, universal plug set (UK/EU Plugset)
Expand
View Pricing
OH30127966
Ohaus
®
Adventurer
®
Precision Balance,
Model AX1502, weighing capacity 1520 g, resolution: 0.01 g, AC/DC input 230 V, universal plug set
Expand
View Pricing
OH30206084
Ohaus
®
Adventurer
®
Precision Balance,
Model AX2201/E, weighing capacity 2200 g, resolution: 0.1 g, AC/DC input 230 V, universal plug set
Expand
View Pricing
OH30127971
Ohaus
®
Adventurer
®
Precision Balance,
Model AX4201/E, weighing capacity 4200 g, resolution: 0.1 g, AC/DC input 230 V, universal plug set
Expand
View Pricing
OH30206080
Ohaus
®
Adventurer
®
Precision Balance,
Model AX422/E, weighing capacity 420 g, resolution: 0.01 g, AC/DC input 230 V, universal plug set
Expand
View Pricing
OH30100623
Ohaus
®
Adventurer
®
Precision Balance,
Model AX223M, weighing capacity 220 g, resolution: 0.001 g, AC/DC input 210 - 2310 V, universal plug set (UK/EU Plugset)
Expand
View Pricing
OH30127962
Ohaus
®
Adventurer
®
Precision Balance,
Model AX423, weighing capacity 420 g, resolution: 0.001 g, AC/DC input 210 - 2310 V, universal plug set (UK/EU Plugset)
Expand
View Pricing
OH30122616
Ohaus
®
Adventurer
®
Precision Balance,
Model AX4202, weighing capacity 4200 g, resolution: 0.01 g, AC/DC input 210 - 2310 V, universal plug set (UK/EU Plugset)
Expand
View Pricing
OH30127967
Ohaus
®
Adventurer
®
Precision Balance,
Model AX1502/E, weighing capacity 1520 g, resolution: 0.01 g, AC/DC input 230 V, universal plug set
Expand
View Pricing
OH30206083
Ohaus
®
Adventurer
®
Precision Balance,
Model AX2201, weighing capacity 2200 g, resolution: 0.1 g, AC/DC input 230 V, universal plug set
Expand
View Pricing
Page 1 of 2
Page 1 of 2