Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Ohaus
®
Adventurer
®
Analytical Balance
Products
Product Category
balances (9)
Manufacturer
Weighing Capacity (g)
Package Size
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
OH30122612
Ohaus
®
Adventurer
®
Analytical Balance,
Model AX224/E, weighing capacity 220 g, resolution: 0.1 mg, AC/DC input 210 - 2310 V, universal plug set (UK/EU Plugset)
Expand
View Pricing
OH30122657
Ohaus
®
Adventurer
®
Analytical Balance,
Model AX124/E, weighing capacity 120 g, resolution: 0.1 mg, AC/DC input 210 - 2310 V, universal plug set (UK/EU Plugset)
Expand
View Pricing
OH30122613
Ohaus
®
Adventurer
®
Analytical Balance,
Model AX324, weighing capacity 320 g, resolution: 0.1 mg, AC/DC input 210 - 2310 V, universal plug set (UK/EU Plugset)
Expand
View Pricing
OH30122611
Ohaus
®
Adventurer
®
Analytical Balance,
Model AX224, weighing capacity 220 g, resolution: 0.1 mg, AC/DC input 210 - 2310 V, universal plug set (UK/EU Plugset)
Expand
View Pricing
OH30122610
Ohaus
®
Adventurer
®
Analytical Balance,
Model AX124, weighing capacity 120 g, resolution: 0.1 mg, AC/DC input 210 - 2310 V, universal plug set (UK/EU Plugset)
Expand
View Pricing
OH30100629
Ohaus
®
Adventurer
®
Analytical Balance,
Model AX5202M, weighing capacity 5200 g, resolution: 0.01 g, AC/DC input 210 - 2310 V, universal plug set (UK/EU Plugset)
Expand
View Pricing
OH30100621
Ohaus
®
Adventurer
®
Analytical Balance,
Model AX224M, weighing capacity 220 g, resolution: 0.1 mg, AC/DC input 210 - 2310 V, universal plug set (UK/EU Plugset)
Expand
View Pricing
OH30100622
Ohaus
®
Adventurer
®
Analytical Balance,
Model AX324M, weighing capacity 320 g, resolution: 0.1 mg, AC/DC input 210 - 2310 V, universal plug set (UK/EU Plugset)
Expand
View Pricing
OH30100620
Ohaus
®
Adventurer
®
Analytical Balance,
Model AX124M, weighing capacity 120 g, resolution: 0.1 mg, AC/DC input 210 - 2310 V, universal plug set (UK/EU Plugset)
Expand
View Pricing