Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Nunclon
®
cell culture dishes
Products
Product Category
dishes (8)
Brand
Sigma-Aldrich (4)
Manufacturer
Binding Type
Capacity (mL)
Diameter (mm)
Height (mm)
Length (mm)
Package Size
Sterility
Sterilization Method
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
D7804
Nunclon
®
cell culture dishes,
diam. × H 35 mm × 10 mm
Expand
View Pricing
Z755923
Nunclon
®
cell culture dishes,
diam. × H 100 mm × 15 mm, sterile; γ-irradiated
Expand
View Pricing
D8054
Nunclon
®
cell culture dishes,
diam. × H 60 mm × 15 mm
Expand
View Pricing
TMO150318
Nunclon
®
cell culture dishes,
diam. 35 mm, with lid, Nunclon
®
Delta treated, case of 500 ea
Expand
View Pricing
D8679
Nunclon
®
cell culture dishes,
L × W × H 245 mm × 245 mm × 20 mm
Expand
View Pricing
P7866
Nunclon
®
cell culture dishes,
diam. × H 60 mm × 15 mm, round vented with lid
Expand
View Pricing
TMO157150
Nunclon
®
cell culture dishes,
diam. 150 mm, with lid, Nunclon
®
Delta treated, case of 120 ea
Expand
View Pricing
TMO174888
Nunclon
®
cell culture dishes,
diam. 60 mm, with lid, Nunclon
®
Delta treated, case of 120 ea
Expand
View Pricing