Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
N-
p
-Tosyl-L-phenylalanine chloromethyl ketone
Synonym(s):
(
S
)-1-Chloro-3-tosylamido-4-phenyl-2-butanone, (
S
)-1-Chloro-4-phenyl-3-tosylamido-2-butanone, TPCK, Tosyl-L-phenylalanyl-chloromethane
Empirical Formula (Hill Notation):
C
17
H
18
ClNO
3
S
CAS Number:
402-71-1
Molecular Weight:
351.85
EC Number:
206-954-6
Products
Brand
Sigma-Aldrich (1)
Biological Source
Color
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
T4376
N-
p
-Tosyl-L-phenylalanine chloromethyl ketone,
powder, ≥97% (TLC)
Expand
View Pricing