Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
n-Pentane
Synonym(s):
Pentane
Linear Formula:
CH
3
(CH
2
)
3
CH
3
CAS Number:
109-66-0
Molecular Weight:
72.15
Products
Product Category
solvents (6)
Brand
Supelco (6)
Boiling Point (°C)
Formula Weight
Physical Form
Product Line
Quality Segment
Technique
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
1.07177
n-Pentane,
for analysis EMSURE
®
Expand
View Pricing
8.20957
n-Pentane,
EMPLURA
®
Expand
View Pricing
1.00882
n-Pentane,
for gas chromatography ECD and FID SupraSolv
®
Expand
View Pricing
1.07288
n-Pentane,
hypergrade for organic trace analysis SupraSolv
®
Expand
View Pricing
1.07179
n-Pentane,
for spectroscopy Uvasol
®
Expand
View Pricing
1.07176
n-Pentane,
about 95%, EMPLURA
®
Expand
View Pricing