Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Norell
®
Standard Series
™
5 mm NMR tubes
Products
Product Category
tubes (24)
Color
Manufacturer
Diameter (mm)
ID (mm)
Length (in)
OD (mm)
Color
Frequency (MHz)
Package Size
Filter Products
Showing 1-20 of 24
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
NOR508UP7
Norell
®
Standard Series
™
5 mm NMR tubes,
frequency 500 MHz
Expand
View Pricing
NOR507HP7
Norell
®
Standard Series
™
5 mm NMR tubes,
frequency 400 MHz
Expand
View Pricing
NOR5027
Norell
®
Standard Series
™
5 mm NMR tubes,
frequency 100 MHz
Expand
View Pricing
NOR507HP8
Norell
®
Standard Series
™
5 mm NMR tubes,
frequency 400 MHz
Expand
View Pricing
NORST5007
Norell
®
Standard Series
™
5 mm NMR tubes,
frequency 100 MHz
Expand
View Pricing
NOR5028
Norell
®
Standard Series
™
5 mm NMR tubes,
frequency 100 MHz
Expand
View Pricing
NOR508UP8
Norell
®
Standard Series
™
5 mm NMR tubes,
frequency 500 MHz
Expand
View Pricing
NORXR557
Norell
®
Standard Series
™
5 mm NMR tubes,
frequency 300 MHz, L 7 in.
Expand
View Pricing
NOR506P7
Norell
®
Standard Series
™
5 mm NMR tubes,
frequency 300+ MHz
Expand
View Pricing
NOR509UP7
Norell
®
Standard Series
™
5 mm NMR tubes,
frequency 600 MHz
Expand
View Pricing
Z690139
Norell
®
Standard Series
™
5 mm NMR tubes,
frequency 100 MHz, L 7 in., orange cap
Expand
View Pricing
NORST5008
Norell
®
Standard Series
™
5 mm NMR tubes,
frequency 100 MHz, L 8 in.
Expand
View Pricing
Z690112
Norell
®
Standard Series
™
5 mm NMR tubes,
frequency 100 MHz, L 7 in., green cap
Expand
View Pricing
Z690104
Norell
®
Standard Series
™
5 mm NMR tubes,
frequency 100 MHz, L 7 in., blue cap
Expand
View Pricing
NOR509UP8
Norell
®
Standard Series
™
5 mm NMR tubes,
frequency 600 MHz
Expand
View Pricing
Z690147
Norell
®
Standard Series
™
5 mm NMR tubes,
frequency 100 MHz, L 7 in., black cap
Expand
View Pricing
NOR506P8
Norell
®
Standard Series
™
5 mm NMR tubes,
frequency 300+ MHz
Expand
View Pricing
Z692123
Norell
®
Standard Series
™
5 mm NMR tubes,
frequency 100 MHz, red cap
Expand
View Pricing
NORXR558
Norell
®
Standard Series
™
5 mm NMR tubes,
frequency 300 MHz, L 8 in.
Expand
View Pricing
NOR505P7
Norell
®
Standard Series
™
5 mm NMR tubes,
frequency 200 MHz
Expand
View Pricing
Page 1 of 2
Page 1 of 2