Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Norell
®
Standard Series
™
10 mm NMR tubes
Products
Product Category
tubes (6)
Manufacturer
Diameter (mm)
ID (mm)
Length (in)
OD (mm)
Color
Frequency (MHz)
Package Size
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
NOR1008UP7
Norell
®
Standard Series
™
10 mm NMR tubes,
frequency 400 MHz
Expand
View Pricing
NOR10017
Norell
®
Standard Series
™
10 mm NMR tubes,
frequency 200 MHz, L 7 in.
Expand
View Pricing
NOR1005P7
Norell
®
Standard Series
™
10 mm NMR tubes,
frequency 300 MHz
Expand
View Pricing
NOR1008UP8
Norell
®
Standard Series
™
10 mm NMR tubes,
frequency 400 MHz
Expand
View Pricing
NOR1005P8
Norell
®
Standard Series
™
10 mm NMR tubes,
frequency 300 MHz
Expand
View Pricing
NOR10018
Norell
®
Standard Series
™
10 mm NMR tubes,
frequency 200 MHz, L 8 in.
Expand
View Pricing