Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Norell
®
Select Series
™
5 mm NMR tubes
Products
Product Category
tubes (14)
Manufacturer
Diameter (mm)
ID (mm)
Length (in)
OD (mm)
Color
Frequency (MHz)
Package Size
Filter Products
Showing 1-14 of 14
Compare
Product Number
Description
Pricing
NORS54007
Norell
®
Select Series
™
5 mm NMR tubes,
frequency 400 MHz
Expand
View Pricing
NORS56007
Norell
®
Select Series
™
5 mm NMR tubes,
frequency 600 MHz
Expand
View Pricing
NORS55007
Norell
®
Select Series
™
5 mm NMR tubes,
frequency 500 MHz
Expand
View Pricing
NORS58007
Norell
®
Select Series
™
5 mm NMR tubes,
frequency 800 MHz
Expand
View Pricing
NORS52007
Norell
®
Select Series
™
5 mm NMR tubes,
frequency 200 MHz, L 7 in.
Expand
View Pricing
NORS54008
Norell
®
Select Series
™
5 mm NMR tubes,
frequency 400 MHz
Expand
View Pricing
NORS53007
Norell
®
Select Series
™
5 mm NMR tubes,
frequency 300 MHz
Expand
View Pricing
NORS56008
Norell
®
Select Series
™
5 mm NMR tubes,
frequency 600 MHz
Expand
View Pricing
NORS55008
Norell
®
Select Series
™
5 mm NMR tubes,
frequency 500 MHz
Expand
View Pricing
NORS52008
Norell
®
Select Series
™
5 mm NMR tubes,
frequency 200 MHz
Expand
View Pricing
NORS59007
Norell
®
Select Series
™
5 mm NMR tubes,
frequency 900 MHz
Expand
View Pricing
NORS59008
Norell
®
Select Series
™
5 mm NMR tubes,
frequency 900 MHz
Expand
View Pricing
NORS53008
Norell
®
Select Series
™
5 mm NMR tubes,
frequency 300 MHz
Expand
View Pricing
NORS58008
Norell
®
Select Series
™
5 mm NMR tubes,
frequency 800 MHz
Expand
View Pricing