Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Nicotine solution
Empirical Formula (Hill Notation):
C
10
H
14
N
2
Molecular Weight:
162.23
Products
Brand
Supelco (7)
Formula Weight
Format
Chemical Composition
Special Grade
analytical standard (6)
Technique
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
612057
Nicotine solution,
NMR reference standard, 2 mg/mL in methanol-d
4
(99.8 atom % D), TMS 0.05 %, NMR tube size 5 mm × 8 in.
Expand
View Pricing
612030
Nicotine solution,
NMR reference standard, 2 mg/mL in DMSO-
d
6
(99.9 atom % D), TMS 0.05 %, NMR tube size 5 mm × 8 in.
Expand
View Pricing
613223
Nicotine solution,
NMR reference standard, 20 mg/mL in acetone-d
6
(99.9 atom % D), TMS 0.05 %, CFCl
3
0.4 %, NMR tube size 5 mm × 8 in.
Expand
View Pricing
612626
Nicotine solution,
NMR reference standard, 20 mg/mL in acetone-d
6
(99.9 atom % D), TMS 0.05 %, NMR tube size 5 mm × 8 in.
Expand
View Pricing
612049
Nicotine solution,
NMR reference standard, 2 mg/mL in acetonitrile-d
3
(99.8 atom % D), TMS 0.05 %, NMR tube size 5 mm × 8 in.
Expand
View Pricing
612014
Nicotine solution,
NMR reference standard, 2 mg/mL in benzene-d
6
(99.6 atom % D), TMS 0.05 %, NMR tube size 5 mm × 8 in.
Expand
View Pricing
612022
Nicotine solution,
NMR reference standard, 2 mg/mL in chloroform-d (99.8 atom % D), TMS 0.05 %, NMR tube size 5 mm × 8 in.
Expand
View Pricing