
Neohesperidin dihydrochalcone impurity B

Synonym(s):
7-[[2-O-(6-Deoxy-α-L-mannopyranosyl)-β-D-glucopyranosyl]oxy]-5-hydroxy-2-(3-hydroxy-4-methoxyphenyl)- 4H-1-benzopyran-4-one, 3′,5,7-Trihydroxy 4′-methoxyflavone 7-neohesperidoside, Diosmetin 7-neohesperidoside, Neodiosmin
Empirical Formula (Hill Notation):
C28H32O15
CAS Number:
Molecular Weight:
608.54
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing