Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Natural PS Liposomes
Products
Brand
Sigma-Aldrich (4)
Biological Source
Tissue
Chemical Composition
Quality Segment
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
CPS-601
Natural PS Liposomes,
Phosphatidylserine (PS)-based - 100% Brain PS (5mM)
Expand
View Pricing
CPS-603
Natural PS Liposomes,
Phosphatidylserine (PS)-based - Brain PS:Cholesterol (70:30 molar ratio 5mM)
Expand
View Pricing
CPS-602
Natural PS Liposomes,
Phosphatidylserine (PS)-based - 100% Soy PS (5mM)
Expand
View Pricing
CPS-604
Natural PS Liposomes,
Phosphatidylserine (PS)-based - Soy PS:Cholesterol (70:30 molar ratio 5mM)
Expand
View Pricing