Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Nalgene
®
wide-mouth bottle
Products
Product Category
bottles (23)
Color
Manufacturer
Capacity (mL)
Package Size
Special Grade
Natural (17)
Filter Products
Showing 1-20 of 23
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
TMO2103-0032
Nalgene
®
wide-mouth bottle,
low-density polyethylene, capacity 1000 mL, natural, case of 24 ea
Expand
View Pricing
TMO2103-0016
Nalgene
®
wide-mouth bottle,
low-density polyethylene, capacity 500 mL, natural, case of 48 ea
Expand
View Pricing
TMO312189-0032
Nalgene
®
wide-mouth bottle,
high-density polyethylene, capacity 1000 mL, natural, case of 50 ea
Expand
View Pricing
TMO312185-0002
Nalgene
®
wide-mouth bottle,
high-density polyethylene, capacity 60 mL, amber, case of 1000 ea
Expand
View Pricing
TMO312189-0002
Nalgene
®
wide-mouth bottle,
high-density polyethylene, capacity 60 mL, natural, case of 1000 ea
Expand
View Pricing
TMO312104-0008
Nalgene
®
wide-mouth bottle,
high-density polyethylene, capacity 250 mL, natural, case of 250 ea
Expand
View Pricing
TMO312189-0008
Nalgene
®
wide-mouth bottle,
high-density polyethylene, capacity 250 mL, natural, case of 250 ea
Expand
View Pricing
TMO312185-0016
Nalgene
®
wide-mouth bottle,
high-density polyethylene, capacity 500 mL, amber, case of 125 ea
Expand
View Pricing
TMO312104-0016
Nalgene
®
wide-mouth bottle,
high-density polyethylene, capacity 500 mL, natural, case of 125 ea
Expand
View Pricing
TMO312185-0032
Nalgene
®
wide-mouth bottle,
high-density polyethylene, capacity 1000 mL, amber, case of 50 ea
Expand
View Pricing
TMO2103-0002
Nalgene
®
wide-mouth bottle,
low-density polyethylene, capacity 60 mL, natural, case of 72 ea
Expand
View Pricing
TMO312189-0016
Nalgene
®
wide-mouth bottle,
high-density polyethylene, capacity 500 mL, natural, case of 125 ea
Expand
View Pricing
TMO312185-0008
Nalgene
®
wide-mouth bottle,
high-density polyethylene, capacity 250 mL, amber, case of 250 ea
Expand
View Pricing
TMO2103-0008
Nalgene
®
wide-mouth bottle,
low-density polyethylene, capacity (250 mL), natural, case of 72 ea
Expand
View Pricing
TMO312185-0004
Nalgene
®
wide-mouth bottle,
high-density polyethylene, capacity 125 mL, amber, case of 500 ea
Expand
View Pricing
TMO2103-0004
Nalgene
®
wide-mouth bottle,
low-density polyethylene, capacity 125 mL, natural, case of 72 ea
Expand
View Pricing
TMO312104-0004
Nalgene
®
wide-mouth bottle,
high-density polyethylene, capacity 125 mL, natural, case of 500 ea
Expand
View Pricing
TMO2103-0001
Nalgene
®
wide-mouth bottle,
low-density polyethylene, capacity 30 mL, natural, case of 72 ea
Expand
View Pricing
TMO312104-0032
Nalgene
®
wide-mouth bottle,
high-density polyethylene, capacity 1000 mL, natural, case of 50 ea
Expand
View Pricing
TMO312189-0001
Nalgene
®
wide-mouth bottle,
high-density polyethylene, capacity 30 mL, natural, case of 1000 ea
Expand
View Pricing
Page 1 of 2
Page 1 of 2