Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Nalgene
®
volumetric flask
Products
Product Category
flasks (10)
Brand
Sigma-Aldrich (4)
Capacity (mL)
Closure Type
Color
Package Size
Sterility
Style
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
Z131245
Nalgene
®
volumetric flask,
volume 50 mL, accuracy: 0.10 mL, translucent polypropylene copolymer (PPCO)
Expand
View Pricing
Z131253
Nalgene
®
volumetric flask,
volume 100 mL, accuracy: 0.16 mL, translucent polypropylene copolymer (PPCO)
Expand
View Pricing
Z131237
Nalgene
®
volumetric flask,
volume 1,000 mL, accuracy: 0.6 mL, transparent polymethylpentene
Expand
View Pricing
Z131296
Nalgene
®
volumetric flask,
volume 1,000 mL, accuracy: 0.60 mL, translucent polypropylene copolymer (PPCO)
Expand
View Pricing
Z131199
Nalgene
®
volumetric flask,
volume 50 mL, accuracy: 0.1 mL, transparent polymethylpentene
Expand
View Pricing
Z131229
Nalgene
®
volumetric flask,
volume 500 mL, accuracy: 0.40 mL, transparent polymethylpentene
Expand
View Pricing
Z131202
Nalgene
®
volumetric flask,
volume 100 mL, accuracy: 0.16 mL, transparent polymethylpentene
Expand
View Pricing
Z131210
Nalgene
®
volumetric flask,
volume 250 mL, accuracy: 0.24 mL, transparent polymethylpentene
Expand
View Pricing
Z131288
Nalgene
®
volumetric flask,
volume 500 mL, accuracy: 0.40 mL, translucent polypropylene copolymer (PPCO)
Expand
View Pricing
Z131261
Nalgene
®
volumetric flask,
volume 250 mL, accuracy: 0.24 mL, translucent polypropylene copolymer (PPCO)
Expand
View Pricing