Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Nalgene
®
straight-sided wide-mouth jar
Products
Manufacturer
Capacity (mL)
Color
Package Size
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
TMO2116-1000
Nalgene
®
straight-sided wide-mouth jar,
polycarbonate, jar capacity 1000 mL, clear, case of 16 ea
Expand
View Pricing
TMO2116-0125
Nalgene
®
straight-sided wide-mouth jar,
polycarbonate, jar capacity 125 mL, clear, case of 24 ea
Expand
View Pricing
TMO2116-0500
Nalgene
®
straight-sided wide-mouth jar,
polycarbonate, clear, case of 16 ea, jar capacity 500 mL
Expand
View Pricing
TMO2116-0250
Nalgene
®
straight-sided wide-mouth jar,
polycarbonate, jar capacity 250 mL, clear, case of 24 ea
Expand
View Pricing
TMO2116-0015
Nalgene
®
straight-sided wide-mouth jar,
polycarbonate, jar capacity 15 mL, clear, case of 48 ea
Expand
View Pricing
TMO2116-0060
Nalgene
®
straight-sided wide-mouth jar,
polycarbonate, jar capacity 60 mL, clear, case of 48 ea
Expand
View Pricing
TMO2116-0030
Nalgene
®
straight-sided wide-mouth jar,
polycarbonate, jar capacity 30 mL, clear, case of 48 ea
Expand
View Pricing