Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Nalgene
®
graduated cylinders
Products
Product Category
cylinders (18)
Manufacturer
Volume (ml)
Package Size
Filter Products
Showing 1-18 of 18
Compare
Product Number
Description
Pricing
Z131121
Nalgene
®
graduated cylinders,
volume 1000 mL, accuracy: 5.0 mL, polypropylene
Expand
View Pricing
TMO3662-4000
Nalgene
®
graduated cylinders,
polypropylene, volume 4000 mL, case of 2 ea
Expand
View Pricing
Z720445
Nalgene
®
graduated cylinders,
volume 2000 mL, accuracy: 12.0 mL, polymethylpentene
Expand
View Pricing
Z131059
Nalgene
®
graduated cylinders,
volume 500 mL, accuracy: 2.6 mL, polymethylpentene
Expand
View Pricing
Z131040
Nalgene
®
graduated cylinders,
volume 100 mL, accuracy: 0.6 mL, polymethylpentene
Expand
View Pricing
Z131105
Nalgene
®
graduated cylinders,
volume 100 mL, accuracy: 0.6 mL, polypropylene
Expand
View Pricing
Z131067
Nalgene
®
graduated cylinders,
volume 1000 mL, accuracy: 5.0 mL, polymethylpentene
Expand
View Pricing
Z131024
Nalgene
®
graduated cylinders,
volume 25 mL, accuracy: 0.3 mL, polymethylpentene
Expand
View Pricing
Z131032
Nalgene
®
graduated cylinders,
volume 50 mL, accuracy: 0.4 mL, polymethylpentene
Expand
View Pricing
Z131016
Nalgene
®
graduated cylinders,
volume 10 mL, accuracy: 0.1 mL, polymethylpentene
Expand
View Pricing
Z131075
Nalgene
®
graduated cylinders,
volume 10 mL, accuracy: 0.1 mL, polypropylene
Expand
View Pricing
Z720453
Nalgene
®
graduated cylinders,
volume 4000 mL, accuracy: 29.0 mL, polymethylpentene
Expand
View Pricing
TMO3662-2000
Nalgene
®
graduated cylinders,
polypropylene, volume 2000 mL, case of 4 ea
Expand
View Pricing
Z131113
Nalgene
®
graduated cylinders,
volume 500 mL, accuracy: 2.6 mL, polypropylene
Expand
View Pricing
Z131083
Nalgene
®
graduated cylinders,
volume 25 mL, accuracy: 0.3 mL, polypropylene
Expand
View Pricing
Z131091
Nalgene
®
graduated cylinders,
volume 50 mL, accuracy: 0.4 mL, polypropylene
Expand
View Pricing
TMO3663-0250
Nalgene
®
graduated cylinders,
PMP, volume 250 mL, case of 12 ea
Expand
View Pricing
TMO3662-0250
Nalgene
®
graduated cylinders,
polypropylene, volume 250 mL, case of 12 ea
Expand
View Pricing