Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Nalgene
®
Dropper Bottle without tip or cap
Products
Product Category
bottles (6)
Manufacturer
Capacity (mL)
Volume (ml)
Color
Package Size
Special Grade
Natural (3)
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
TMO312751-9025
Nalgene
®
Dropper Bottle without tip or cap,
white low-density polyethylene, volume 8 mL, case of 2000 ea
Expand
View Pricing
TMO312750-9050
Nalgene
®
Dropper Bottle without tip or cap,
natural low-density polyethylene, volume 15 mL, case of 2000 ea
Expand
View Pricing
TMO312750-9125
Nalgene
®
Dropper Bottle without tip or cap,
natural low-density polyethylene, volume 4 mL, case of 2000 ea
Expand
View Pricing
TMO312751-9050
Nalgene
®
Dropper Bottle without tip or cap,
white low-density polyethylene, volume 15 mL, case of 2000 ea
Expand
View Pricing
TMO312750-9025
Nalgene
®
Dropper Bottle without tip or cap,
natural low-density polyethylene, volume 8 mL, case of 2000 ea
Expand
View Pricing
TMO312751-9125
Nalgene
®
Dropper Bottle without tip or cap,
white low-density polyethylene, volume 4 mL, case of 2000 ea
Expand
View Pricing