Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Nalgene
®
bottles, style 2006
Products
Product Category
bottles (6)
Brand
Sigma-Aldrich (2)
Capacity (mL)
Diameter (mm)
Cap
Bottle material
Color
Vial Bottle Material
Vial Bottle Color
Package Size
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
B8657
Nalgene
®
bottles, style 2006,
capacity 1,000 mL
Expand
View Pricing
B8157
Nalgene
®
bottles, style 2006,
capacity 60 mL
Expand
View Pricing
B8532
Nalgene
®
bottles, style 2006,
capacity 500 mL
Expand
View Pricing
B8407
Nalgene
®
bottles, style 2006,
capacity 250 mL
Expand
View Pricing
B8282
Nalgene
®
bottles, style 2006,
capacity 125 mL
Expand
View Pricing
B8032
Nalgene
®
bottles, style 2006,
capacity 30 mL
Expand
View Pricing