Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Nalgene
®
baffled shake flask
Products
Product Category
flasks (4)
Brand
Sigma-Aldrich (4)
Binding Type
Capacity (mL)
Diameter (mm)
Working Volume (ml)
Cap
Color
Package Size
Sterility
Style
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
F0527
Nalgene
®
baffled shake flask,
capacity 2,000 mL, Beaded neck
Expand
View Pricing
F0152
Nalgene
®
baffled shake flask,
flask capacity 250 mL, Beaded neck
Expand
View Pricing
F0277
Nalgene
®
baffled shake flask,
flask capacity 500 mL, Straight neck
Expand
View Pricing
F0402
Nalgene
®
baffled shake flask,
flask capacity 1,000 mL, Straight neck
Expand
View Pricing