
Nafcillin sodium salt monohydrate

Synonym(s):
6-(2-Ethoxy-1-naphthamido)penicillin
Linear Formula:
C21H21N2O5SNa · H2O
CAS Number:
Molecular Weight:
454.47
Products
Product Category
Brand
Formula Weight
Physical Form
Activity Spectrum
Quality Segment
Mode of Action
Showing 1-1 of 1
Compare
Product Number
Description
Pricing