Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
N3-PEG-NHS
Synonym(s):
N3-PEG-NHS
Linear Formula:
N
3
CH
2
CH
2
O(CH
2
CH
2
O)nCH
2
COOC
4
H
4
O
2
N
Products
Brand
Sigma-Aldrich (2)
Physical Form
Functionality
Shape
Polymer Composition
Polymer Type
Reaction Type
Reactivity
Reagent Type
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
JKA5086
N3-PEG-NHS,
average M
N
3,500, cross-linking reagent click chemistry amine reactive
Expand
View Pricing
JKA5088
N3-PEG-NHS,
average M
N
5,000, cross-linking reagent click chemistry amine reactive
Expand
View Pricing