Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
N
-(1-Naphthyl)ethylenediamine dihydrochloride
Synonym(s):
2-(1-Naphthylamino)ethylamine dihydrochloride, N-(1-Naphthyl)ethylenediamine dihydrochloride
Linear Formula:
C
10
H
7
NHCH
2
CH
2
NH
2
· 2HCl
CAS Number:
1465-25-4
Molecular Weight:
259.17
EC Number:
215-981-2
Products
Product Category
building blocks (2)
Brand
Sigma-Aldrich (2)
Supelco (1)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Special Grade
ACS reagent (2)
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
222488
N
-(1-Naphthyl)ethylenediamine dihydrochloride,
ACS reagent, >98%
Expand
View Pricing
N9125
N
-(1-Naphthyl)ethylenediamine dihydrochloride,
≥98%
Expand
View Pricing
1.06237
N
-(1-Naphthyl)ethylenediamine dihydrochloride,
GR for analysis ACS
Expand
View Pricing
33461
N
-(1-Naphthyl)ethylenediamine dihydrochloride,
for determination of sulfonamide and nitrite, ACS reagent, ≥98%
Expand
View Pricing