Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
m
-Xylene
Synonym(s):
1,3-Dimethylbenzene, m-Xylene,
m
-Xylene
Linear Formula:
C
6
H
4
(CH
3
)
2
CAS Number:
108-38-3
Molecular Weight:
106.17
EC Number:
203-576-3
Products
Product Category
solvents (3)
Brand
Sigma-Aldrich (4)
Supelco (1)
Boiling Point (°C)
Formula Weight
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Reference Material Type
Special Grade
Anhydrous (1)
Puriss (1)
Puriss P.A. (1)
analytical standard (1)
certified reference material (1)
pharmaceutical secondary standard (1)
Technique
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
185566
m
-Xylene,
ReagentPlus
®
, 99%
Expand
View Pricing
95672
m
-Xylene,
puriss. p.a., ≥99.0% (GC)
Expand
View Pricing
8.22337
m
-Xylene,
for synthesis
Expand
View Pricing
5.89592
m
-Xylene,
Expand
View Pricing
95670
m
-Xylene,
analytical standard
Expand
View Pricing
PHR1313
m
-Xylene,
Pharmaceutical Secondary Standard; Certified Reference Material
Expand
View Pricing