Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Muscovite Mica for AFM
Synonym(s):
Muscovite Mica for AFM
Products
Brand
Sigma-Aldrich (3)
Physical Form
Diameter (mm)
Package Size
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
AFM-71855-15-10
Muscovite Mica for AFM,
(V-1 Quality), size 15 mm × 15 mm, pack of 10
Expand
View Pricing
AFM-71856-02
Muscovite Mica for AFM,
(V-1 Quality), diam. 12 mm, pack of 10
Expand
View Pricing
AFM-71856-03
Muscovite Mica for AFM,
(V-1 Quality), size 15 mm, pack of 10
Expand
View Pricing