Monoclonal Anti-CD11b−FITC antibody produced in mouse
Synonym(s):
Monoclonal Anti-CD11b, Anti-ITGAM, Anti-Mac-1 α
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Physical Form
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-2 of 2