Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Mineral Oil Rotational Viscosity Standard
Products
Manufacturer
Format
Application
Reference Material Type
Special Grade
certified reference material (35)
Filter Products
Showing 1-20 of 36
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
RTM4
Mineral Oil Rotational Viscosity Standard,
viscosity 2.144 mPa.s (25.00 °C)
Expand
View Pricing
RTM11
Mineral Oil Rotational Viscosity Standard,
viscosity 29.04 mPa.s (25.00 °C)
Expand
View Pricing
RTM8
Mineral Oil Rotational Viscosity Standard,
viscosity 10.37 mPa.s (25.00 °C)
Expand
View Pricing
RTM35
Mineral Oil Rotational Viscosity Standard,
viscosity 10030 mPa.s (25.00 °C)
Expand
View Pricing
RTM15
Mineral Oil Rotational Viscosity Standard,
viscosity 150.0 mPa.s (25.00 °C)
Expand
View Pricing
RTM24
Mineral Oil Rotational Viscosity Standard,
viscosity 1008 mPa.s (25.00 °C)
Expand
View Pricing
RTM3
Mineral Oil Rotational Viscosity Standard,
viscosity 0.9525 mPa.s (25.00 °C)
Expand
View Pricing
RTM21
Mineral Oil Rotational Viscosity Standard,
viscosity 494.0 mPa.s (25.00 °C)
Expand
View Pricing
RTM29
Mineral Oil Rotational Viscosity Standard,
viscosity 4024 mPa.s (25.00 °C)
Expand
View Pricing
RTM20
Mineral Oil Rotational Viscosity Standard,
viscosity 389.1 mPa.s (25.00 °C)
Expand
View Pricing
RTM13
Mineral Oil Rotational Viscosity Standard,
viscosity 75.19 mPa.s (25.00 °C)
Expand
View Pricing
RTM18
Mineral Oil Rotational Viscosity Standard,
viscosity 300.8 mPa.s (25.00 °C)
Expand
View Pricing
RTM7
Mineral Oil Rotational Viscosity Standard,
viscosity 7.520 mPa.s (25.00 °C)
Expand
View Pricing
RTM39
Mineral Oil Rotational Viscosity Standard,
viscosity 72328 mPa.s (25.00 °C)
Expand
View Pricing
RTM12
Mineral Oil Rotational Viscosity Standard,
viscosity 55.68 mPa.s (25.00 °C)
Expand
View Pricing
RTM17
Mineral Oil Rotational Viscosity Standard,
viscosity 250.0 mPa.s (25.00 °C)
Expand
View Pricing
RTM31
Mineral Oil Rotational Viscosity Standard,
viscosity 5738 mPa.s (25.00 °C)
Expand
View Pricing
RTM6
Mineral Oil Rotational Viscosity Standard,
viscosity 4.745 mPa.s (25.00 °C)
Expand
View Pricing
RTM38
Mineral Oil Rotational Viscosity Standard,
viscosity 40049 mPa.s (25.00 °C)
Expand
View Pricing
RTM1
Mineral Oil Rotational Viscosity Standard,
viscosity 0.3051 mPa.s (25.00 °C)
Expand
View Pricing
Page 1 of 2
Page 1 of 2