Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Micro spatulas
Products
Product Category
spatulas/scoops (9)
Brand
Sigma-Aldrich (5)
Length (in)
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
Z193216
Micro spatulas,
one scraper/spoon end, L 6 1/2 in. (165 mm)
Expand
View Pricing
Z513334
Micro spatulas,
flat rounded/tapered PTFE coated ends, L 7 5/8 in. (190 mm)
Expand
View Pricing
Z513342
Micro spatulas,
flat rounded ends, L 8 in. (203 mm)
Expand
View Pricing
Z513393
Micro spatulas,
oval spoon/plastic handle, L 6 1/2 in. (165 mm)
Expand
View Pricing
Z513385
Micro spatulas,
flat rounded/tapered ends, L 7 in. (178 mm)
Expand
View Pricing
Z513377
Micro spatulas,
flat rounded/tapered ends, L 12 in. (305 mm)
Expand
View Pricing
Z513350
Micro spatulas,
flat rounded ends, L 12 in. (305 mm)
Expand
View Pricing
Z193208
Micro spatulas,
Curved at one end with knob at the other end, L 5 1/12 in. (140 mm)
Expand
View Pricing
Z513369
Micro spatulas,
flat rounded/tapered ends, L 8 in. (203 mm)
Expand
View Pricing