Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Microfil
®
funnel with S-Pak
®
Products
Product Category
funnels (13)
Brand
Millipore (13)
Agency
Diameter (mm)
Compatibility
Hydrophobicity
Application
Color
Pore size (µm)
Sterility
Sterilization Method
Technique
Filter Products
Showing 1-13 of 13
Compare
Product Number
Description
Pricing
MIHAWG100
Microfil
®
funnel with S-Pak
®
,
sample volume 100 mL, mixed cellulose esters (MCE), pore size 0.45 μm, filter diam. 47 mm, white filter, gridded filter
Expand
View Pricing
MIHAWG250
Microfil
®
funnel with S-Pak
®
,
sample volume 250 mL, mixed cellulose esters (MCE), pore size 0.45 μm, filter diam. 47 mm, white filter, gridded filter
Expand
View Pricing
MIHVWP100
Microfil
®
funnel with S-Pak
®
,
100 mL Funnel volume with S-Pak PVDF membrane, 0.45 μm pore size, 47 mm diameter, white plain filter
Expand
View Pricing
MIHABG250
Microfil
®
funnel with S-Pak
®
,
sample volume 250 mL, mixed cellulose esters (MCE), pore size 0.45 μm, filter diam. 47 mm, black filter, gridded filter
Expand
View Pricing
MIRAWG250
Microfil
®
funnel with S-Pak
®
,
sample volume 250 mL, mixed cellulose esters (MCE), pore size 1.2 μm, filter diam. 47 mm, white filter, gridded filter, To detect yeast or mold in difficult-to-filter sample
Expand
View Pricing
MIHABG100
Microfil
®
funnel with S-Pak
®
,
sample volume 100 mL, mixed cellulose esters (MCE), pore size 0.45 μm, filter diam. 47 mm, black filter, gridded filter
Expand
View Pricing
MIGSWG250
Microfil
®
funnel with S-Pak
®
,
sample volume 250 mL, mixed cellulose esters (MCE), pore size 0.22 μm, filter diam. 47 mm, white filter, gridded filter, for water, beverage, and pharmaceutical application.
Expand
View Pricing
MIGSWG100
Microfil
®
funnel with S-Pak
®
,
sample volume 100 mL, mixed cellulose esters (MCE), pore size 0.22 μm, filter diam. 47 mm, white filter, gridded filter
Expand
View Pricing
MIAABG100
Microfil
®
funnel with S-Pak
®
,
sample volume 100 mL, mixed cellulose esters (MCE), pore size 0.8 μm, filter diam. 47 mm, black filter, gridded filter, To detect yeast and mold
Expand
View Pricing
MIAAWG100
Microfil
®
funnel with S-Pak
®
,
sample volume 100 mL, mixed cellulose esters (MCE), pore size 0.8 μm, filter diam. 47 mm, white filter, gridded filter, To detect yeast and mold
Expand
View Pricing
MIHCWG250
Microfil
®
funnel with S-Pak
®
,
sample volume 250 mL, mixed cellulose esters (MCE), pore size 0.7 μm, filter diam. 47 mm, white filter, gridded filter
Expand
View Pricing
MIRAWG100
Microfil
®
funnel with S-Pak
®
,
sample volume 100 mL, mixed cellulose esters (MCE), pore size 1.2 μm, filter diam. 47 mm, white filter, gridded filter, To detect yeast or mold in difficult-to-filter samples
Expand
View Pricing
MIHCWG100
Microfil
®
funnel with S-Pak
®
,
sample volume 100 mL, mixed cellulose esters (MCE), pore size 0.7 μm, filter diam. 47 mm, white filter, gridded filter
Expand
View Pricing