Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Microcentrifuge Tube
Products
Product Category
tubes (5)
Manufacturer
Capacity (mL)
Diameter (mm)
Cap
Color
Package Size
Special Grade
Natural (1)
Sterility
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
HS4323
Microcentrifuge Tube,
natural polypropylene, capacity 1.5 mL, pack of 500 ea
Expand
View Pricing
HS4323K
Microcentrifuge Tube,
tube capacity 1.5 mL, pack of 500 ea
Expand
View Pricing
HS4325
Microcentrifuge Tube,
conical bottom, capacity 1.5 mL, pack of 1000 ea
Expand
View Pricing
HS4422
Microcentrifuge Tube,
capacity 0.5 mL, pack of 1000 ea
Expand
View Pricing
HS4423
Microcentrifuge Tube,
capacity 0.2 mL, pack of 1000 ea
Expand
View Pricing