Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
MG-132
Synonym(s):
Z-Leu-Leu-Leu-al, MG-132
Empirical Formula (Hill Notation):
C
26
H
41
N
3
O
5
CAS Number:
133407-82-6
Molecular Weight:
475.62
Products
Product Category
bioactive small molecules (3)
Brand
Sigma-Aldrich (3)
Color
Formula Weight
Physical Form
Research Area
Quality Segment
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
474790
MG-132,
A cell-permeable, potent, reversible proteasome inhibitor (K
i
= 4 nM).
Expand
View Pricing
474791
MG-132,
InSolution, ≥98%, 10 mM, reversible proteasome inhibitor
Expand
View Pricing
474787
MG-132,
≥95% by HPLC, Potent, reversible, and cell-permeable proteasome inhibitor (Kᵢ = 4 nM).
Expand
View Pricing