Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Mettler-Toledo electrolyte for reference electrodes
Products
Manufacturer
Physical Form
Volume (ml)
Filter Products
Showing 1-11 of 11
Compare
Product Number
Description
Pricing
MT51350072
Mettler-Toledo electrolyte for reference electrodes,
3 mol/L KCl, volume 250 mL
Expand
View Pricing
MT51350076
Mettler-Toledo electrolyte for reference electrodes,
Friscolyt-B, volume 250 mL
Expand
View Pricing
MT51343180
Mettler-Toledo electrolyte for reference electrodes,
3 mol/L KCl, volume 25 mL
Expand
View Pricing
MT51350088
Mettler-Toledo electrolyte for reference electrodes,
LiCl 1M in EtOH, volume 6 x 30 mL
Expand
View Pricing
MT51350074
Mettler-Toledo electrolyte for reference electrodes,
3 mol/L KCl Sat AgCl, volume 250 mL
Expand
View Pricing
MT51350080
Mettler-Toledo electrolyte for reference electrodes,
3 mol/L KCl, volume 6 x 250 mL
Expand
View Pricing
MT51340294
Mettler-Toledo electrolyte for reference electrodes,
InLab
®
605, volume 25 mL
Expand
View Pricing
MT51343184
Mettler-Toledo electrolyte for reference electrodes,
3 mol/L KCl Sat AgCl, volume 25 mL
Expand
View Pricing
MT51343185
Mettler-Toledo electrolyte for reference electrodes,
Friscolyt-B, volume 25 mL
Expand
View Pricing
MT51350084
Mettler-Toledo electrolyte for reference electrodes,
Friscolyt-B, volume 6 x 250 mL
Expand
View Pricing
MT51350082
Mettler-Toledo electrolyte for reference electrodes,
3 mol/L KCl Sat AgCl, volume 6 x 250 mL
Expand
View Pricing