Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Mettler-Toledo conductivity standard
Products
Manufacturer
Physical Form
Volume (ml)
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
MT51302153
Mettler-Toledo conductivity standard,
Conductivity: 84 μs/cm, volume 250 mL
Expand
View Pricing
MT51300169
Mettler-Toledo conductivity standard,
Conductivity: 10 μs/cm, volume 250 mL
Expand
View Pricing
MT51350092
Mettler-Toledo conductivity standard,
Conductivity: 1413 μs/cm, volume 250 mL
Expand
View Pricing
MT51350094
Mettler-Toledo conductivity standard,
Conductivity: 12.88 mS, volume 250 mL
Expand
View Pricing
MT51300170
Mettler-Toledo conductivity standard,
Conductivity: 500 μs/cm, volume 250 mL
Expand
View Pricing
MT51302050
Mettler-Toledo conductivity standard,
Conductivity: 12.88 mS/cm, volume 30 x 20 mL , sachets
Expand
View Pricing
MT51302049
Mettler-Toledo conductivity standard,
Conductivity: 1413 μs/cm, volume 30 x 20 mL , sachets
Expand
View Pricing
MT51350096
Mettler-Toledo conductivity standard,
Conductivity: 1413 μs/cm, volume 6 x 250 mL
Expand
View Pricing
MT51350098
Mettler-Toledo conductivity standard,
Conductivity: 12.88 mS, volume 6 x 250 mL
Expand
View Pricing