
Methyl 4-iodosalicylate

Synonym(s):
Methyl 2-hydroxy-4-iodobenzoate
Empirical Formula (Hill Notation):
C8H7IO3
CAS Number:
Molecular Weight:
278.04
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing