Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Methyl 4-hydroxybenzoate sodium salt
Synonym(s):
Methyl 4-hydroxybenzoate sodium salt, Sodium methyl 4-hydroxybenzoate
Linear Formula:
Na
+
[4-(O)C
6
H
4
(COOCH
3
)]
-
CAS Number:
5026-62-0
Molecular Weight:
174.13
EC Number:
225-714-1
Products
Product Category
hormones (1)
preservatives (1)
salts (1)
Brand
SAFC (1)
Sigma-Aldrich (1)
Supelco (1)
Formula Weight
Manufacturer
Physical Form
Product Line
Agency
Quality Segment
Application
Reference Material Type
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
1.06756
Methyl 4-hydroxybenzoate sodium salt,
EMPROVE
®
ESSENTIAL, Ph. Eur., BP, NF
Expand
View Pricing
PHR9290
Methyl 4-hydroxybenzoate sodium salt,
certified reference material,
Trace
CERT
®
, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Expand
View Pricing
85265
Methyl 4-hydroxybenzoate sodium salt,
tested according to Ph. Eur.
Expand
View Pricing