
Methyl 4-(Acetylamino)-2-hydroxybenzoate

Synonym(s):
Methyl 4-(acetylamino)salicylate, Methyl 4-acetamido-2-hydroxybenzoate
Empirical Formula (Hill Notation):
C10H11NO4
CAS Number:
Molecular Weight:
209.20
Products
Brand
Formula Weight
Physical Form
Agency
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing