Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Mega Rack
®
tube rack
Products
Product Category
racks (10)
Color
Manufacturer
Height (mm)
Length (mm)
Color
Package Size
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
HS120835
Mega Rack
®
tube rack,
blue polypropylene
Expand
View Pricing
HS120114
Mega Rack
®
tube rack,
double, for 13-16 mm tubes, white
Expand
View Pricing
HS120834
Mega Rack
®
tube rack,
white polypropylene
Expand
View Pricing
HS24322E
Mega Rack
®
tube rack,
single, for 10-13 mm tubes, white
Expand
View Pricing
HS24323E
Mega Rack
®
tube rack,
single, for 13-16 mm tubes, white
Expand
View Pricing
HS24323A
Mega Rack
®
tube rack,
single, for 13-16 mm tubes, blue
Expand
View Pricing
HS120115
Mega Rack
®
tube rack,
double, for 13-16 mm tubes, blue
Expand
View Pricing
HS24322A
Mega Rack
®
tube rack,
single, for 10-13 mm tubes, blue
Expand
View Pricing
HS120113
Mega Rack
®
tube rack,
double, for 10-13 mm tubes, blue
Expand
View Pricing
HS120112
Mega Rack
®
tube rack,
double, for 10-13 mm tubes, white
Expand
View Pricing